missing translation for 'onlineSavingsMsg'
Learn More

Wheaton™ 10 μL Inoculation Loop

Inoculation Loop for delivering a known volume of fluid sample to enable accurate microbiological analysis

Brand:  Wheaton™ 711300

204.27 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 16415009

  • £424.32 / Pack of 6000
Estimated Shipment: 23-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For dispersing samples on agar surface
  • Meet quantitative standards for management of microbiological samples
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Single-use
ABS
Short
6000
Not Autoclavable
199 mm
Sterile
Blue
Packaged in peel pouch
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Wheaton™ 10 μL Inoculation Loop > Size: Short

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.