Learn More
SI Analytics™ 10 mL Exchangeable Unit
SI Analytics™ 10 mL Exchangeable Unit is an accessory for TitroLine™ and Titronic™ models.
Brand:  SI Analytics™ 285220310
Product Code. 13485898
  • £700.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With integrated chip for reagent data and brown glass bottle for titrant
  • Includes GL 45 and S 40-bottle adapter
TRUSTED_SUSTAINABILITY
Specifications
Specifications
WA 10
Integrated chip, glass bottle, bottle adapter, drip tubes, and titration itp
10 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SI Analytics™ 10 mL Exchangeable Unit > Capacity: 10 mL