missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCR2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems FAB151N-100
381.60 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
CCR2 Monoclonal specifically detects CCR2 in Human samples. It is validated for Flow Cytometry.
Specifications
CCR2 | |
Monoclonal | |
Alexa Fluor 700 | |
NP_001116868 | |
CCR2 | |
NS0 mouse myeloma cell line transfected with human CCR2 Met1-Leu360 Accession # NP_001116868 | |
100 Tests | |
729230 | |
Human | |
Purified |
Flow Cytometry | |
48607 | |
Flow Cytometry 5 uL/10^6 cells | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
Mouse | |
Protein A or G purified from ascites | |
Primary | |
Detects human CCR2 transfectants but not the parental cell line or CCR5 transfectants. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human CCR2 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™ > 100 Tests; Alexa Fluor 700
Spot an opportunity for improvement?Share a Content Correction