Learn More
Thermo Scientific™ 13mm Autosampler Vial Screw Thread Caps
13mm Screw Thread Caps feature a 13-425 thread finish. Available in an assortment of styles, pre-assembled caps and septa minimize contamination.
£5.38 - £387.00
Product Code | Brand | Color | Septum | Material | Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Septum | Material | Type | Price | Quantity & Availability | ||||||
12410188
|
Thermo Scientific™
C4015-1A |
Black | None | Polypropylene | Open Top Screw Cap |
£5.38
Pack of 100 |
In Stock
Log in to see stock. |
||||||
12430188
|
Thermo Scientific™
C4015-1W |
White | None | Polypropylene | Open Top Screw Cap | N/A | N/A | N/A | |||||
12400228
|
Thermo Scientific™
C4015-66 |
Black | None | Phenolic | Open Top Screw Cap |
£78.10
Pack of 1000 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
12410228
|
Thermo Scientific™
C4015-66A |
Black, Red, White | Red PTFE/White Silicone | Phenolic, PTFE, Silicone | Open Top Screw Cap |
£54.85
Pack of 100 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
12420228
|
Thermo Scientific™
C4015-67A |
Black, Red, White | PTFE/Silicone, Bonded | Polypropylene, PTFE, Silicone | Open Top Screw Cap | N/A | N/A | N/A | |||||
12075540
|
Thermo Scientific™
C4015-30A |
Black, Ivory, Red | Ivory PTFE/Red Rubber | Polypropylene, PTFE, Rubber | Open Top Screw Cap |
£29.38
Pack of 100 |
In Stock
Log in to see stock. |
||||||
12410198
|
Thermo Scientific™
C4015-40A |
Black, Red, White | PTFE/Silicone/PTFE | Polypropylene, PTFE, Silicone | Open Top Screw Cap |
£40.35
Pack of 100 |
Estimated Shipment: 29-05-2025
Log in to see stock. |
||||||
10476854
|
Thermo Scientific™
C4015-45W |
White, Red, White | Red PTFE/White Silicone, Soft | Polypropylene, PTFE, Silicone | Open Top Screw Cap | N/A | N/A | N/A | |||||
12470208
|
Thermo Scientific™
C4015-55BLK |
Black, Blue, White | PTFE/Silicone, Pre-slit | Polypropylene, PTFE, Silicone | Open Top Screw Cap |
£44.55
Pack of 100 |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
10497234
|
Thermo Scientific™
C4015-55W |
White, Blue, White | PTFE/Silicone, Pre-slit | Polypropylene, PTFE, Silicone | Open Top Screw Cap | N/A | N/A | N/A | |||||
10027684
|
Thermo Scientific™
C4015-75A |
Black, Red, White | Red PTFE/White Silicone | Polypropylene, PTFE, Silicone | Open Top Screw Cap | N/A | N/A | N/A | |||||
12460228
|
Thermo Scientific™
C4015-75W |
White, Red, White | Red PTFE/White Silicone | Polypropylene, PTFE, Silicone | Open Top Screw Cap | N/A | N/A | N/A | |||||
12490208
|
Thermo Scientific™
C4015-5A |
Natural | Integral Molded | Polypropylene, Polyethylene | Screw Cap |
£12.50
Pack of 100 |
Estimated Shipment: 05-06-2025
Log in to see stock. |
||||||
11538250
|
Thermo Scientific™
B7815-13 |
White | PTFE/PE Foam Liner | Urethane | Screw Cap |
£35.15
Pack of 100 |
Estimated Shipment: 27-05-2025
Log in to see stock. |
||||||
11593690
|
Thermo Scientific™
12-SC |
Black | None | Polypropylene | Screw Cap |
£126.00
Pack of 500 |
In Stock
Log in to see stock. |
||||||
11573710
|
Thermo Scientific™
12-SCS |
Black | None (no hole) | Polypropylene | Screw Cap |
£56.25
Pack of 500 |
Estimated Shipment: 02-06-2025
Log in to see stock. |
||||||
11503710
|
Thermo Scientific™
12-SC(Y) |
Yellow | None | Polypropylene | Open Top Screw Cap | N/A | N/A | N/A | |||||
10295712
|
Thermo Scientific™
13-SC-ST15 |
Black, White, Red | Silicone/PTFE | Polypropylene, PTFE, Silicone | Open Top Screw Cap | N/A | N/A | N/A | |||||
10514673
|
Thermo Scientific™
13-SCST |
White | PTFE | Urea | Screw Cap | N/A | N/A | N/A | |||||
11553710
|
Thermo Scientific™
12-SC-8RT1 |
Black, Clear, Red | Natural Rubber/PTFE | Polypropylene, PTFE, Rubber | Screw Cap |
£208.04
Pack of 500 |
Estimated Shipment: 09-06-2025
Log in to see stock. |
||||||
11563710
|
Thermo Scientific™
12-SC-ST2 |
Black, Red, White | PTFE/Silicone | Polypropylene, PTFE, Silicone | Screw Cap | N/A | N/A | N/A | |||||
Description
Thermo Scientific™ 13mm Screw Thread Caps feature a 13-425 thread finish and are available in an assortment of styles to match your HPLC, GC or IC application needs. Convenient pre-assembled caps and septa minimize contamination from handling. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.
Autosampler Vial Screw Thread CapsLimited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.