Learn More
Keystone Cleanroom Products 1421B Tyvek™ Autoclavable Bowl Cover, with Steam Indicator
1421B Tyvek™ Autoclavable Bowl Covers prevent microbial and particulate contamination to critical parts.
£1390.00 - £4725.00
Specifications
Product Type | Cover |
---|---|
Autoclavable | Autoclavable |
Closure Type | None |
Color | White |
Disposable | Single-use |
Product Code | Brand | Width (English) | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (English) | Width (Metric) | Price | Quantity & Availability | |||||
30020149
![]() |
Keystone Cleanroom Products
BCTK20 |
20 in. | 51 cm |
£3005.00
Pack of 500 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020100
![]() |
Keystone Cleanroom Products
BCTK24 |
24 in. | 61 cm |
£3235.00
Pack of 500 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020131
![]() |
Keystone Cleanroom Products
BCTK26 |
26 in. | 66 cm |
£3890.00
Pack of 500 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020188
![]() |
Keystone Cleanroom Products
BCTK30 |
30 in. | 76 cm |
£4725.00
Pack of 500 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020115
![]() |
Keystone Cleanroom Products
BCTK32-2PLY8 |
32 in. | 81.3 cm |
£1815.00
Pack of 100 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
30020233
![]() |
Keystone Cleanroom Products
BCTK36 |
36 in. | 91 cm |
£1855.00
Pack of 100 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020353
![]() |
Keystone Cleanroom Products
BCTK38 |
38 in. | 97 cm |
£1990.00
Pack of 100 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020388
![]() |
Keystone Cleanroom Products
BCTK40 |
40 in. | 102 cm |
£1390.00
Pack of 50 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
30020092
![]() |
Keystone Cleanroom Products
BCTK42 |
42 in. | 107 cm |
£1545.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020383
![]() |
Keystone Cleanroom Products
BCTK44 |
44 in. | 112 cm |
£1720.00
Pack of 50 |
Estimated Shipment: 17-07-2025
Log in to see stock. |
|||||
30020085
![]() |
Keystone Cleanroom Products
BCTK46 |
46 in. | 117 cm |
£1720.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020174
![]() |
Keystone Cleanroom Products
BCTK48 |
48 in. | 122 cm |
£2090.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020004
![]() |
Keystone Cleanroom Products
BCTK50 |
50 in. | 127 cm |
£2200.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020019
![]() |
Keystone Cleanroom Products
BCTK52-2PLY12 |
52 in. | 132.1 cm |
£3675.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020196
![]() |
Keystone Cleanroom Products
BCTK54 |
54 in. | 137 cm |
£2310.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020156
![]() |
Keystone Cleanroom Products
BCTK56 |
56 in. | 142 cm |
£2440.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30020328
![]() |
Keystone Cleanroom Products
BCTK60 |
60 in. | 152 cm |
£2570.00
Pack of 50 |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
30019996
![]() |
Keystone Cleanroom Products
BCTK72 |
72 in. | 183 cm |
£3190.00
Pack of 50 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
30020034
![]() |
Keystone Cleanroom Products
BCTK90 |
90 in. | 229 cm |
£3675.00
Pack of 50 |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
Description
- Prevent microbial and particulate contamination to critical parts, while the fill line set-up activities are performed
- Manufactured and packaged in a controlled cleanroom environment
- Reduces labor costs
- Simplifies the conformance to wrapping SOP's
- Steam Sterilization Indicator - Autoclavable at 121°C to 125°C
- Tear and puncture resistant
- Hydrophobic
- Lint-free
Specifications
Cover | |
None | |
Single-use | |
Pharmaceutical Equipment and Components | |
Non-sterile | |
Up to 250°F | |
5.3 mil |
Autoclavable | |
White | |
Steam Sterilization | |
Tyvek | |
0.13 mm | |
Up to 125°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Keystone Cleanroom Products 1421B Tyvek™ Autoclavable Bowl Cover, with Steam Indicator