Learn More
Fisherbrand™ 150 Homogenizer Package
Includes everything necessary to process samples as small as 200μL to 100mL. Fisherbrand™ 150 Homogenizer Package includes the 150w motor as well as accessories such as 2 saw tooth stainless steel generator probes, disposable generator probes and more.
Brand: Fisherbrand™ 15595809
449.68 GBP valid until 2025-05-18
Use promo code "24096" to get your promotional price.
Description
- Processes laboratory samples, from 200μL to 100mL
- Powerful 150w motor
- Lightweight ergonomic design
- Quiet operation, under 72db
- Available with disposable generator probes (adapter for use is included)
- Cross-contamination can be completely eliminated with the use of plastic generator probes
- Stainless steel generator probes are recommended for larger and tougher samples
Certifications
CE
Services
One year

Specifications
5000 to 35,000 rpm | |
200 μL to 100 mL | |
16.5 cm | |
150 W motor, Flat bottom Stainless Steel generator probe (5 x 75 mm), Saw tooth Stainless Steel generator probe (7 x 115 mm), Saw tooth Stainless Steel generator probe (10 x 115 mm), Stand clamp, Motor stand assembly, Disposable probe adapter, Disposable generator probes, Instruction manual and Tool kit | |
UK Plug |
220V UK Cord, 150W | |
5.1 cm | |
Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics | |
1 Ea. | |
466 g |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
7Hours
17Minutes
56Seconds
42
Your input is important to us. Please complete this form to provide feedback related to the content on this product.