missing translation for 'onlineSavingsMsg'
Learn More
1H-Pyrazolo[3,4-b]pyridine, 97%, Thermo Scientific Chemicals Product Code.: 15448278

1H-Pyrazolo[3,4-b]pyridine, 97%, Thermo Scientific Chemicals

Product Code. 15448278
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15448278

Brand: Thermo Scientific Alfa Aesar H61710.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 271-73-8
Molecular Formula C6H5N3
Molecular Weight (g/mol) 119.127
MDL Number MFCD05663981
InChI Key GVLRTOYGRNLSDW-UHFFFAOYSA-N
Synonym 1h-pyrazolo 3,4-b pyridine, 2h-pyrazolo 3,4-b pyridine, 7-azaindazole, pyrazolo 3,4-b pyridine, 7-aza-1h-indazole, pyrazolo 5,4-b pyridine, 7-aza indazole, 7-azabenzpyrazole, acmc-1cs3b, 2h pyrazolo 3,4 b pyridinShow MoreShow Less
PubChem CID 2755850
IUPAC Name 1H-pyrazolo[3,4-b]pyridine
SMILES C1=CC2=C(NN=C2)N=C1

Specifications

Quantity 1 g
Formula Weight 119.12
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1H-Pyrazolo[3,4-b]pyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81300195 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61710.MD,H61710.03
Lot NumberN01D027 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH61710.MD,H61710.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1H-Pyrazolo[3,4-b]pyridine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.