missing translation for 'onlineSavingsMsg'
Learn More
2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals Product Code.: 10722402

2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals

Product Code. 10722402
1 kg, Glass bottle
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10722402

Brand: Thermo Scientific Acros 413900010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-65-6
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00038500
InChI Key LLHKCFNBLRBOGN-UHFFFAOYSA-N
Synonym 1-methoxy-2-propyl acetate, pgmea, 2-acetoxy-1-methoxypropane, propylene glycol monomethyl ether acetate, propylene glycol methyl ether acetate, 1-methoxy-2-acetoxypropane, 2-propanol, 1-methoxy-, acetate, 2-methoxy-1-methylethyl acetate, propyleneglycol monomethyl ether acetate, acetic acid, 2-methoxy-1-methylethyl esterShow MoreShow Less
PubChem CID 7946
IUPAC Name 1-methoxypropan-2-yl acetate
SMILES CC(COC)OC(=O)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -87.0°C
Color Colorless
Density 0.9600g/mL
Boiling Point 146.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CH(CH3)OC(O)CH3
Refractive Index 1.4010 to 1.4030
Quantity 1 kg
UN Number 1993
Specific Gravity 0.96
Solubility Information Solubility in water: 19.8g/L (25°C)
Formula Weight 132.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(1-Methoxy)propyl acetate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465664 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog Number413900010,413900050
Lot NumberA0448755 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number413900010,413900050
Lot NumberA0455562 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number413900010,413900050
Lot NumberA0455561 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number413900010,413900050
Lot NumberA0399698 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number413900010,413900050
compliance-icons
Product Identifier
  • 2-(1-Methoxy)propyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(1-Methoxy)propyl acetate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.