missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%, Thermo Scientific Chemicals

£27.54 - £164.82

Chemical Identifiers

CAS 2997-92-4
Molecular Formula C8H18N6·2HCl
Molecular Weight (g/mol) 271.19
MDL Number MFCD00142725
InChI Key LXEKPEMOWBOYRF-UHFFFAOYSA-N
Synonym aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap
PubChem CID 76344
IUPAC Name 2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride
SMILES CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10218502
View Documents
Thermo Scientific Acros
401560250
25 g Plastic bottle
£27.54
25g
Please to purchase this item. Need a web account? Register with us today!
10005650
View Documents Promotion Details
Thermo Scientific Acros
401560500
50 g Plastic Bottle
£46.43
50g
Please to purchase this item. Need a web account? Register with us today!
10698414
View Documents
Thermo Scientific Acros
401562500
250 g Plastic bottle
£164.82
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
Specifications

Chemical Identifiers

2997-92-4
271.19
LXEKPEMOWBOYRF-UHFFFAOYSA-N
76344
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl
C8H18N6·2HCl
MFCD00142725
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride

Specifications

2997-92-4
100.0
0.4200g/mL
Authentic
C8H18N6·2HCl
04,IV,3379
Solubility in water: soluble.
CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl
271.19
271.19
Granules
97.5
White
1% max.
97.5 to 102.5% (on dry substance) (Argentometry)
MFCD00142725
aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap
LXEKPEMOWBOYRF-UHFFFAOYSA-N
2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride
76344
98%
2,2′-Azobis(2-methylpropionamidine) dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Self-heating; may catch fire.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Keep cool.
Protect from sunlight.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/at

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 221-070-

Product Identifier
  • 2,2''-Azobis(2-methylpropionamidine) dihydrochloride

Signal Word
  • Danger

Hazard Category
  • Self-heating substance Category 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H251-Self-heating: may catch fire.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only