Learn More
2,2'-Bipyridine, 99+%, Thermo Scientific Chemicals
Description
Reagent for iron determinations.2,2'-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reagent for iron determinations.2,2′-bipyridine acts as a bidentate chelating ligand which form complexes with transition metal ions and shows antifungal, antibacterial and antiviral activity. It is used for the colorimetric determination of iron as well as oxidation reduction indicators to confirm the presence of ferrous ion in soils. It forms complexes with ruthenium and platinum, exhibit intense luminescence, which may have practical applications. Copper(I) bipyridine complexes involved in the oxidation of alcohols under aerobic conditions.
Solubility
Soluble in organic solvents and slightly soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from sources of ignition.

Specifications
Specifications
Melting Point | 70°C |
Boiling Point | 272°C to 273°C |
Flash Point | 121°C (249°F) |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 113089 |
Merck Index | 14,3347 |
Solubility Information | Soluble in organic solvents and slightly soluble in water. |
IUPAC Name | 2,2'-bipyridine |
Formula Weight | 156.19 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10251845 | Certificate of Analysis | 02/08/2024 | A15782.22,A15782.14,A15782.36 |
10202289 | Certificate of Analysis | 23/11/2021 | A15782.22,A15782.14,A15782.36 |
10212143 | Certificate of Analysis | 23/11/2021 | A15782.22,A15782.14,A15782.36 |
10193717 | Certificate of Analysis | 23/11/2021 | A15782.22,A15782.14,A15782.36 |
J15Z001 | Certificate of Analysis | 23/11/2021 | A15782.22,A15782.14,A15782.36 |
Safety and Handling

- 2,2'-Bipyridine
- Danger
- Acute toxicity Category 3
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 2,2'-Bipyridine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.