missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,4-Trifluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
CAS Min % | 97.0 |
Density | 1.6400g/mL |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | F3C6H2SO2Cl |
Quantity | 1 g |
IUPAC Name | 2,3,4-trifluorobenzenesulfonyl chloride |
Formula Weight | 230.59 |
Percent Purity | 97% |
Chemical Name or Material | 2, 3, 4-Trifluorobenzene-1-sulfonyl chloride |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,3,4-Trifluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction