missing translation for 'onlineSavingsMsg'
Learn More
2,3,4-Trihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals Product Code.: 11424784

2,3,4-Trihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 11424784
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11424784

Brand: Thermo Scientific Alfa Aesar A13745.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2144-08-3

Specifications

Melting Point 159°C to 163°C
Molecular Formula C8H9BO2
MDL Number MFCD00239441
Quantity 5 g
Beilstein 2328658
Sensitivity Air Sensitive
Synonym 4-vinylphenylboronic acid, 4-vinylbenzeneboronic acid, 4-vinylphenyl boronic acid, 4-ethenylphenyl boronic acid, 4-vinylboronic acid, p-styrylboronic acid, p-vinylphenylboronic acid, styrene-4-boronic acid, boronic acid, 4-ethenylphenyl, 4-styreneboronic acid
InChI Key QWMJEUJXWVZSAG-UHFFFAOYSA-N
SMILES OB(O)C1=CC=C(C=C)C=C1
IUPAC Name (4-ethenylphenyl)boronic acid
Molecular Weight (g/mol) 147.97
PubChem CID 2734393
Formula Weight 154.12
Percent Purity 98%
Chemical Name or Material 2,3,4-Trihydroxybenzaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252489 Certificate TypeCertificate of Analysis Date30/09/2024 Catalog NumberA1374501,A13745.06,A13745.03,A13745.14
Lot Number10194468 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog NumberA1374501,A13745.06,A13745.03,A13745.14
Lot Number10097844 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA1374501,A13745.06,A13745.14
Lot Number10194468 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA1374501,A13745.06,A13745.14
Lot NumberD5033B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA1374501,A13745.06,A13745.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,3,4-Trihydroxybenzaldehyde, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.