missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,3-Dimethylphenoxy)acetic acid, 98%, Thermo Scientific™
£10.18 - £22.96
Chemical Identifiers
CAS | 2935-63-9 |
---|---|
Molecular Formula | C10H12O3 |
Molecular Weight (g/mol) | 180.20 |
MDL Number | MFCD00016814 |
InChI Key | AVDBMBYECMTQJQ-UHFFFAOYSA-N |
Synonym | 2,3-dimethylphenoxyacetic acid, 2-2,3-dimethylphenoxy acetic acid, 2,3-dimethylphenoxy acetic acid, 2,3-xylyloxyacetic acid, acmc-20aog1, enamine_005510, 2,3-dimethylphenoxyessig, cambridge id 5161587, 2,3-dimethylphenoxy acetic acid, acetic acid, 2,3-dimethylphenoxy |
PubChem CID | 76239 |
IUPAC Name | 2-(2,3-dimethylphenoxy)acetic acid |
SMILES | CC1=CC=CC(OCC(O)=O)=C1C |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
2935-63-9 | |
180.20 | |
AVDBMBYECMTQJQ-UHFFFAOYSA-N | |
76239 | |
CC1=CC=CC(OCC(O)=O)=C1C |
C10H12O3 | |
MFCD00016814 | |
2,3-dimethylphenoxyacetic acid, 2-2,3-dimethylphenoxy acetic acid, 2,3-dimethylphenoxy acetic acid, 2,3-xylyloxyacetic acid, acmc-20aog1, enamine_005510, 2,3-dimethylphenoxyessig, cambridge id 5161587, 2,3-dimethylphenoxy acetic acid, acetic acid, 2,3-dimethylphenoxy | |
2-(2,3-dimethylphenoxy)acetic acid |
Specifications
2935-63-9 | |
97.5% min. (HPLC) | |
(CH3)2C6H3OCH2CO2H | |
2,3-dimethylphenoxyacetic acid, 2-2,3-dimethylphenoxy acetic acid, 2,3-dimethylphenoxy acetic acid, 2,3-xylyloxyacetic acid, acmc-20aog1, enamine_005510, 2,3-dimethylphenoxyessig, cambridge id 5161587, 2,3-dimethylphenoxy acetic acid, acetic acid, 2,3-dimethylphenoxy | |
AVDBMBYECMTQJQ-UHFFFAOYSA-N | |
2-(2,3-dimethylphenoxy)acetic acid | |
76239 | |
98% |
186°C to 190°C | |
C10H12O3 | |
MFCD00016814 | |
Solubility in water: insoluble | |
CC1=CC=CC(OCC(O)=O)=C1C | |
180.20 | |
180.2 | |
(2, 3-Dimethylphenoxy)acetic acid |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 220-911-9