missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-trifluoro-3-methoxybenzoyl chloride, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
Specifications
Assay Percent Range | 95% |
Quantity | 10 g |
Formula Weight | 224.56 |
Percent Purity | 97% |
Chemical Name or Material | 2, 4, 5-Trifluoro-3-methoxybenzoyl chloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,4,5-trifluoro-3-methoxybenzoyl chloride, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction