Learn More
2,4-Dichloropyrimidine, 98+%, Thermo Scientific Chemicals
Description
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,4-Dichloropyrimidine is used in the synthesis of medicinally important 4-aryl-5-pyrimidinylimidazoles. It undergoes effective one-pot, regioselective double Suzuki coupling reaction to yield diarylated pyrimidines.
Solubility
Soluble in water (partly), methanol, chloroform, and ethyl acetate.
Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | 57°C to 62°C |
Boiling Point | 101°C (23 mmHg) |
Quantity | 50 g |
Beilstein | 110911 |
Sensitivity | Moisture sensitive |
Synonym | pyrimidine, 2,4-dichloro, 2,6-dichloropyrimidine, 2,4-dichloro-pyrimidine, 2,4-dichlorpyrimidin, 2,4-dichloro pyrimidine, pyrimidine,2,4-dichloro, pyrimidine,4-dichloro, hydrochloride, zlchem 210, 2,4dichloropyrimidine |
Solubility Information | Soluble in water (partly),methanol,chloroform,and ethyl acetate. |
Formula Weight | 148.98 |
Percent Purity | ≥98% |
Chemical Name or Material | 2,4-Dichloropyrimidine |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10250732 | Certificate of Analysis | 30/08/2024 | A15131.30,A15131.09,A15131.18,A15131.06 |
10246744 | Certificate of Analysis | 24/05/2023 | A15131.30,A15131.09,A15131.18,A15131.06 |
G1512B | Certificate of Analysis | 22/11/2021 | A15131.30,A15131.09,A15131.18,A15131.06 |
10100644 | Certificate of Analysis | 22/11/2021 | A15131.30,A15131.09,A15131.18,A15131.06 |
J3935A | Certificate of Analysis | 22/11/2021 | A15131.30,A15131.09,A15131.18,A15131.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.