missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dimercapto-1,3,4-thiadiazole, 97% (dry wt.), water <3%, Thermo Scientific Chemicals Product Code.: 11400443

2,5-Dimercapto-1,3,4-thiadiazole, 97% (dry wt.), water <3%, Thermo Scientific Chemicals

Product Code. 11400443
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11400443

Brand: Thermo Scientific Alfa Aesar A12615.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimercapto-1,3,4-thiadiazole is used in synthesizing polymers and heavy metal and basic salts. It acts as an additive in lubricating oils, greases and in electrode compositions. It also acts as an intermediate for pharmaceuticals and dyes, as a chelating agent in the analysis of metals. It also serves as an anticorrosion and antirust agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-71-5
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
MDL Number MFCD00003103
InChI Key BIGYLAKFCGVRAN-UHFFFAOYSA-N
Synonym 1,3,4-thiadiazole-2,5-dithiol, 2,5-dimercapto-1,3,4-thiadiazole, bismuthiol i, dimercaptothiadiazole, bismuththiol, 2,5-dimercaptothiadiazole, usaf a-8354, bismuthiol, py 61h, bismuththiol iShow MoreShow Less
PubChem CID 2723630
IUPAC Name 1,3,4-thiadiazolidine-2,5-dithione
SMILES S=C1NNC(=S)S1

Specifications

Melting Point ∼162°C (decomposition)
Quantity 250 g
Beilstein 113444
Solubility Information Soluble in water.
Formula Weight 150.24
Percent Purity 98%
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW17E025 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12615.30,A12615.18
Lot Number10099389 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12615.30,A12615.18
Lot NumberU08F023 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12615.30,A12615.18
Lot Number10033565 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12615.30,A12615.18
Lot Number10185970 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12615.30,A12615.18
compliance-icons
Product Identifier
  • 2,5-Dimercapto-1,3,4-thiadiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1,3,4-Thiadiazolidine-2,5-dithione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimercapto-1,3,4-thiadiazole, 97% (dry wt.), water <3%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.