missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dichloroindophenol, sodium salt hydrate, 90+%, Thermo Scientific Chemicals Product Code.: 13505050

2,6-Dichloroindophenol, sodium salt hydrate, 90+%, Thermo Scientific Chemicals

Product Code. 13505050
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 13505050

Brand: Thermo Scientific Acros 450700250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1266615-56-8
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key CVSUAFOWIXUYQA-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt dihydrate, 2,6-dichloro-4-4-hydroxyphenyl azamethylene cyclohexa-2,5-dien-1-one, oxamet hane, oxamethane, sodium salt, c12h6cl2no2.na.2h2o, 2,6-dichlorophenol-indophenol sodium salt dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino phenolate dihydrate, sodium 4-3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene amino benzenolate dihydrateShow MoreShow Less
PubChem CID 23696612
IUPAC Name sodium;4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]phenolate;dihydrate
SMILES [Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1

Specifications

Assay Percent Range 90+%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Formula Weight 290.07
Percent Purity 90+%
Chemical Name or Material 2, 6-Dichloroindophenol, sodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468520 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number450700250,450701000,450700050
Lot NumberA0450643 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number450700250,450701000,450700050
Lot NumberA0447053 Certificate TypeCertificate of Analysis Date05/12/2022 Catalog Number450700250,450701000,450700050
Lot NumberA0440245 Certificate TypeCertificate of Analysis Date23/03/2022 Catalog Number450700250,450701000,450700050
Lot NumberA0430762 Certificate TypeCertificate of Analysis Date14/06/2021 Catalog Number450700250,450701000,450700050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dichloroindophenol, sodium salt hydrate, 90+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.