Learn More
Chemical Identifiers
CAS | 108-48-5 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.16 |
MDL Number | MFCD00006345 |
InChI Key | OISVCGZHLKNMSJ-UHFFFAOYSA-N |
Synonym | 2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene |
PubChem CID | 7937 |
ChEBI | CHEBI:32548 |
IUPAC Name | 2,6-dimethylpyridine |
SMILES | CC1=CC=CC(C)=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10532331
|
Thermo Scientific Acros
125192500 |
250 mL | Glass bottle |
£36.38
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10686512
|
Thermo Scientific Acros
125190010 |
1 L | Glass bottle |
£97.08
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731354
|
Thermo Scientific Acros
125190025 |
2.5 L | Glass bottle |
£219.09
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-48-5 | |
107.16 | |
OISVCGZHLKNMSJ-UHFFFAOYSA-N | |
7937 | |
2,6-dimethylpyridine |
C7H9N | |
MFCD00006345 | |
2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene | |
CHEBI:32548 | |
CC1=CC=CC(C)=N1 |
Specifications
108-48-5 | |
100.0 | |
0.9200g/mL | |
33°C | |
98.5% min. (GC) | |
1.4966 to 1.4986 | |
20, 244 | |
15, 5677 | |
2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutidene | |
OISVCGZHLKNMSJ-UHFFFAOYSA-N | |
2,6-dimethylpyridine | |
7937 | |
107.15 | |
Liquid |
98.5 | |
-6.0°C | |
143.0°C to 145.0°C | |
Authentic | |
C7H9N | |
MFCD00006345 | |
01,626 | |
0.92 | |
Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol and ether,miscible with dimethylformamide and,tetrahydrofuran | |
CC1=CC=CC(C)=N1 | |
107.16 | |
CHEBI:32548 | |
99% | |
2, 6-Lutidine, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Flammable liquid and vapor.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-587-3
missing translation for 'rtecsNumber' : OK9700000
missing translation for 'tsca' : TSCA
Recommended Storage : This substance may take up color on standing
- 2,6-Lutidine
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P270-Do not eat, drink or smoke when using this product.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only