missing translation for 'onlineSavingsMsg'
Learn More
2,6-Lutidine, 99%, Thermo Scientific Chemicals Product Code.: 10532331

2,6-Lutidine, 99%, Thermo Scientific Chemicals

Product Code. 10532331
250 mL, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10532331

Brand: Thermo Scientific Acros 125192500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-48-5
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.16
MDL Number MFCD00006345
InChI Key OISVCGZHLKNMSJ-UHFFFAOYSA-N
Synonym 2,6-lutidine, lutidine, pyridine, 2,6-dimethyl, 2,6-dimethypyridine, alpha,alpha'-lutidine, alpha,alpha'-dimethylpyridine, 2,6-dimethyl-pyridine, hsdb 79, unii-15fq5d0t3p, 2,6-lutideneShow MoreShow Less
PubChem CID 7937
ChEBI CHEBI:32548
IUPAC Name 2,6-dimethylpyridine
SMILES CC1=CC=CC(C)=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -6.0°C
Density 0.9200g/mL
Boiling Point 143.0°C to 145.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4966 to 1.4986
Quantity 250 mL
Beilstein 20, 244
Fieser 01,626
Merck Index 15, 5677
Specific Gravity 0.92
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: soluble in alcohol and ether,miscible with dimethylformamide and,tetrahydrofuran
Formula Weight 107.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2, 6-Lutidine, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467280 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number125192500,125190025,125190010
Lot NumberA0467279 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number125192500,125190025,125190010
Lot NumberA0466186 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog Number125192500,125190025,125190010
Lot NumberA0463898 Certificate TypeCertificate of Analysis Date20/06/2024 Catalog Number125192500,125190025,125190010
Lot NumberA0458829 Certificate TypeCertificate of Analysis Date19/12/2023 Catalog Number125192500,125190025,125190010
compliance-icons compliance-icons
Product Identifier
  • 2,6-Lutidine
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P270-Do not eat, drink or smoke when using this product.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Lutidine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.