missing translation for 'onlineSavingsMsg'
Learn More
(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals Product Code.: 10549073

(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals

Product Code. 10549073
250 mg, Glass bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10549073

Brand: Thermo Scientific Acros 291632500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2041-14-7
Molecular Formula C2H8NO3P
Molecular Weight (g/mol) 125.06
MDL Number MFCD00008182
InChI Key QQVDJLLNRSOCEL-UHFFFAOYSA-N
Synonym 2-aminoethyl phosphonic acid, ciliatine, phosphonoethylamine, aminoethylphosphonic acid, 2-aminoethane phosphonic acid, 2-aminoethanephosphonic acid, 2-aminoethylphosphonate, beta-aminoethylphosphonic acid, 2-aminoethyl phosphonate, phosphonic acid, 2-aminoethylShow MoreShow Less
PubChem CID 339
ChEBI CHEBI:15573
IUPAC Name 2-aminoethylphosphonic acid
SMILES NCCP(O)(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 296.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NH2CH2CH2P(O)(OH)2
Quantity 250 mg
Beilstein 04, 1792
Solubility Information Solubility in water: soluble in water. Other solubilities: insoluble in methanol and acetone
Formula Weight 125.06
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (2-Aminoethyl)phosphonic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443175 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number291632500,291630010
Lot NumberA0403593 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number291632500,291630010
Lot NumberA0399220 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number291632500,291630010
Lot NumberA0443175 Certificate TypeCertificate of Analysis Date24/06/2022 Catalog Number291632500,291630010
Lot NumberA0412956 Certificate TypeCertificate of Analysis Date11/04/2022 Catalog Number291632500,291630010
compliance-icons
Product Identifier
  • (2-Aminoethyl)phosphonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(2-Aminoethyl)phosphonic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.