missing translation for 'onlineSavingsMsg'
Learn More
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals Product Code.: 11495887

2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals

Product Code. 11495887
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11495887

Brand: Thermo Scientific Alfa Aesar B24441.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10312-55-7
Molecular Formula C8H5NO4
Molecular Weight (g/mol) 179.13
MDL Number MFCD00134536
InChI Key GPNNOCMCNFXRAO-UHFFFAOYSA-L
Synonym aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751Show MoreShow Less
PubChem CID 2724822
IUPAC Name 2-aminoterephthalic acid
SMILES NC1=CC(=CC=C1C([O-])=O)C([O-])=O

Specifications

Melting Point >300°C
Odor Odorless
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 181.15
Percent Purity 99%
Chemical Name or Material 2-Aminoterephthalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249286 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog NumberB24441.22,B24441.06,B24441.14
Lot Number10244307 Certificate TypeCertificate of Analysis Date27/10/2022 Catalog NumberB24441.22,B24441.06,B24441.14
Lot Number10240246 Certificate TypeCertificate of Analysis Date21/04/2022 Catalog NumberB24441.22,B24441.06,B24441.14
Lot NumberG6232A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24441.22,B24441.06,B24441.14
Lot NumberK09Y017 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24441.22,B24441.06,B24441.14
compliance-icons
Product Identifier
  • 2-Aminoterephthalic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2-Aminoterephthalicacid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.