missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals Product Code.: 11994501

2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals

Product Code. 11994501
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11994501

Brand: Thermo Scientific Acros 434460010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1514-82-5
Molecular Formula C3H2BrF3
Molecular Weight (g/mol) 174.95
MDL Number MFCD00077469
InChI Key QKBKGNDTLQFSEU-UHFFFAOYSA-N
Synonym 2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropeneShow MoreShow Less
PubChem CID 272696
IUPAC Name 2-bromo-3,3,3-trifluoroprop-1-ene
SMILES FC(F)(F)C(Br)=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.6500g/mL
Boiling Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3520 to 1.3550 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.65
Formula Weight 174.95
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromo-3, 3, 3-trifluoro-1-propene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435971 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog Number434460050,434460010
Lot NumberA0405868 Certificate TypeCertificate of Analysis Date21/02/2022 Catalog Number434460050,434460010
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-3,3,3-trifluoro-1-propene
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.