Learn More
2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals
£36.20 - £461.00
Chemical Identifiers
CAS | 1451-82-7 |
---|---|
Molecular Formula | C10H11BrO |
Molecular Weight (g/mol) | 227.10 |
MDL Number | MFCD11131402 |
InChI Key | OZLUPIIIHOOPNQ-UHFFFAOYNA-N |
Synonym | 2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone |
PubChem CID | 2734063 |
IUPAC Name | 2-bromo-1-(4-methylphenyl)propan-1-one |
SMILES | CC(Br)C(=O)C1=CC=C(C)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15442279
|
Thermo Scientific Alfa Aesar
|
1 g |
£36.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452279
|
Thermo Scientific Alfa Aesar
|
5 g |
£103.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15462279
|
Thermo Scientific Alfa Aesar
H64881.14 |
25 g |
£461.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.
Solubility
Soluble in chloroform and methanol.
Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Chemical Identifiers
1451-82-7 | |
227.10 | |
OZLUPIIIHOOPNQ-UHFFFAOYNA-N | |
2734063 | |
CC(Br)C(=O)C1=CC=C(C)C=C1 |
C10H11BrO | |
MFCD11131402 | |
2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone | |
2-bromo-1-(4-methylphenyl)propan-1-one |
Specifications
1451-82-7 | |
MFCD11131402 | |
2-bromo-1-p-tolyl propan-1-one, 2-bromo-4'-methylpropiophenone, 2-bromo-1-4-methylphenyl propan-1-one, 2-bromo-4-methylpropiophenone, 2-bromo-1-4-methylphenyl-1-propanone, 1-propanone, 2-bromo-1-4-methylphenyl, 2-bromo-1-p-tolylpropan-1-one, ksc917e0p, 2-bromo-1-p-tolyl-propan-1-one, 4'-methyl-alpha-bromopropiophenone | |
OZLUPIIIHOOPNQ-UHFFFAOYNA-N | |
2-bromo-1-(4-methylphenyl)propan-1-one | |
2734063 | |
98% | |
2-Bromo-4'-methylpropiophenone |
C10H11BrO | |
2207628 | |
Soluble in chloroform and methanol. | |
CC(Br)C(=O)C1=CC=C(C)C=C1 | |
227.10 | |
227.1 | |
Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only