missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™ Product Code.: 11305555

2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11305555
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11305555

Brand: Thermo Scientific Alfa Aesar L19712.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54403-98-4
Molecular Formula C7H3BrClF3O2S
Molecular Weight (g/mol) 323.508
MDL Number MFCD03094394
InChI Key WWEGTYZLWBOTQW-UHFFFAOYSA-N
Synonym 2-bromo-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-bromo-4-trifluoromethyl benzenesulfonyl chloride, 2-bromo-4-trifluoromethyl benzene sulphonyl chloride, 2-bromo-4-trifluoromethyl benzenesulfonylchloride, 2-bromo-4-trifluoromethyl-benzenesulfonyl chloride, 2-bromo-4-trifluoromethyl benzenesulphonyl chloride, benzenesulfonyl chloride, 2-bromo-4-trifluoromethyl, pubchem11716, ksc269c3j, ablock ab-13-6537Show MoreShow Less
PubChem CID 2778611
IUPAC Name 2-bromo-4-(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=CC(=C(C=C1C(F)(F)F)Br)S(=O)(=O)Cl

Specifications

Density 1.865
Boiling Point 235°C to 236°C
Refractive Index 1.527
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 323.52
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-4-(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.