missing translation for 'onlineSavingsMsg'
Learn More
2-Bromopropionic acid, 98%, Thermo Scientific Chemicals Product Code.: 11488197

2-Bromopropionic acid, 98%, Thermo Scientific Chemicals

Product Code. 11488197
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy

Product Code. 11488197

Brand: Thermo Scientific Alfa Aesar A17569.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is employed as pharmaceutical and agrochemical intermediate. It is used as an alkylation agent for mercaptans and other sulfur containing compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as pharmaceutical and agrochemical intermediate. It is used as an alkylation agent for mercaptans and other sulfur containing compounds.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Specifications

CAS 598-72-1
Melting Point 25°C
Density 1.7
Boiling Point 202°C to 203°C
Flash Point 100°C (212°F)
Molecular Formula C3H5BrO2
Refractive Index 1.476
MDL Number MFCD00004211,MFCD00145205
Quantity 250 g
UN Number UN3261
Beilstein 605451
Synonym 2-bromopropionic acid, propanoic acid, 2-bromo, dl-2-bromopropionic, propionic acid, 2-bromo, alpha-bromopropionic acid, dl-2-bromopropionic acid, 2-bromo-propionic acid, .alpha.-bromopropionic acid, propanoic acid, bromo, propanoicacid, 2-bromo
Solubility Information Soluble in water.
InChI Key MONMFXREYOKQTI-UHFFFAOYNA-N
SMILES CC(Br)C(O)=O
IUPAC Name 2-bromopropanoic acid
Molecular Weight (g/mol) 152.98
PubChem CID 11729
Formula Weight 152.98
Percent Purity 98%
Chemical Name or Material 2-Bromopropionic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242460 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog NumberA17569.30
Lot NumberGBFA033172 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17569.30
Lot Number10201213 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17569.30
Lot NumberJ7492A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17569.30
Lot Number10233917 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17569.30
compliance-icons compliance-icons
Product Identifier
  • 2-Bromopropionic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Bromopropionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromopropionic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.