missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haen
Click to view available options
Quantity:
100 mL
1 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency

Specifications
Specifications
Melting Point | -87°C (literature) |
CAS Max % | 100.0000% |
Density | 0.805 g/mL at 25 °C(lit.) |
Boiling Point | 79°C to 81°C |
Quantity | 1 L |
Flash Point | −4°C |
Packaging | Glass Bottle |
Linear Formula | C2H5COCH3 |
UN Number | UN1193 |
Beilstein | 741880 |
Show More |
Safety and Handling


Product Identifier
- 2-Butanone
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : butanone
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Butanone, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haen >
Spot an opportunity for improvement?Share a Content Correction