missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-1-methylpyridinium iodide, 97%, Thermo Scientific Chemicals Product Code.: 11485193

2-Chloro-1-methylpyridinium iodide, 97%, Thermo Scientific Chemicals

Product Code. 11485193
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11485193

Brand: Thermo Scientific Alfa Aesar A12820.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1-methylpyridinium iodide is a reagent in a wide variety of dehydrative coupling reactions. It is also used to form carboxylate esters from acids and alcohols, carboxamides from acids and amines, lactones from -hydroxy acids and carbodiimides from N,N-disubstituted thioureas.

Solubility
Soluble in Methanol (50 mg/ml). Insoluble in water.

Notes
Store at 4°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14338-32-0
Molecular Formula C6H7ClIN
Molecular Weight (g/mol) 255.48
MDL Number MFCD00011984
InChI Key ABFPKTQEQNICFT-UHFFFAOYSA-M
Synonym 2-chloro-1-methylpyridinium iodide, 2-chloro-1-methylpyridin-1-ium iodide, pyridinium, 2-chloro-1-methyl-, iodide, unii-rlq88nyz68, rlq88nyz68, 2-chloro-1-methylpyridiniumiodide, 2-chloro-1-methyl-pyridinium iodide, pubchem16659, acmc-1brmd, ksc494o1lShow MoreShow Less
PubChem CID 167069
IUPAC Name 2-chloro-1-methylpyridin-1-ium;iodide
SMILES [I-].C[N+]1=CC=CC=C1Cl

Specifications

Melting Point ∼200°C (decomposition)
Quantity 25 g
Beilstein 3572320
Sensitivity Light and moisture sensitive
Merck Index 14,6301
Solubility Information Soluble in Methanol (50mg/ml). Insoluble in water.
Formula Weight 255.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-1-methylpyridinium iodide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243710 Certificate TypeCertificate of Analysis Date01/11/2022 Catalog NumberA12820.14,A12820.36,A12820.22
Lot Number10239420 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12820.14,A12820.36,A12820.22
Lot Number10232105 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12820.14,A12820.36,A12820.22
Lot Number10192571 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12820.14,A12820.36,A12820.22
Lot Number10056109 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12820.14,A12820.36,A12820.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-1-methylpyridinium iodide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.