missing translation for 'onlineSavingsMsg'
Learn More
2-Hexanol, 99%, Thermo Scientific Chemicals Product Code.: 10300214

2-Hexanol, 99%, Thermo Scientific Chemicals

Product Code. 10300214
250 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10300214

Brand: Thermo Scientific Acros 156292500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-93-7
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004585
InChI Key QNVRIHYSUZMSGM-UHFFFAOYSA-N
Synonym 2-hexanol, sec-hexyl alcohol, 2-hydroxyhexane, 2-hexyl alcohol, n-butylmethylcarbinol, 2-hexanol, r, 2-hexanol, s, n-c4h9ch oh ch3, methylamyl alcohol, sec-hexanolShow MoreShow Less
PubChem CID 12297
IUPAC Name hexan-2-ol
SMILES CCCCC(C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -23.0°C
Color Colorless
Density 0.8100g/mL
Boiling Point 139.0°C
Flash Point 41°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CH(OH)CH3
Refractive Index 1.4125 to 1.4155
Quantity 250 g
Beilstein 01, 408
Specific Gravity 0.81
Solubility Information Solubility in water: insoluble.
Formula Weight 102.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Hexanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463411 Certificate TypeCertificate of Analysis Date06/06/2024 Catalog Number156292500,156290500,156290100
Lot NumberA0462193 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number156292500,156290500,156290100
Lot NumberA0425919 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number156292500,156290500,156290100
Lot NumberA0451443 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number156292500,156290500,156290100
Lot NumberA0417223 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number156292500,156290500,156290100
compliance-icons compliance-icons
Product Identifier
  • 2-Hexanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Hexanol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.