missing translation for 'onlineSavingsMsg'
Learn More
2-Imidazolidone hemihydrate, 99+%, Thermo Scientific Chemicals Product Code.: 10132540

2-Imidazolidone hemihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 10132540
500 g
Click to view available options
Quantity:
25 g
500 g
2.5 kg
10 kg
This item is not returnable. View return policy

Product Code. 10132540

Brand: Thermo Scientific Acros 122045000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121325-67-5, 120-93-4
Molecular Formula C6H14N4O3
Molecular Weight (g/mol) 190.20
MDL Number MFCD02662355
InChI Key KPRJGGOOWATRNT-UHFFFAOYSA-N
Synonym imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrateShow MoreShow Less
PubChem CID 2723651
IUPAC Name imidazolidin-2-one;hydrate
SMILES O.O=C1NCCN1.O=C1NCCN1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 58.0°C
Color White to Yellow
Density 265g/mL
Boiling Point >100.0°C
Flash Point 265°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Merck Index 14, 4914
Solubility Information Solubility in water: 950g/L (20°C). Other solubilities: 1800g/L water (40°C)
Formula Weight 95.1
Percent Purity 99+%
Physical Form Adhering Crystals or Crystalline Powder
Chemical Name or Material 2-Imidazolidone hemihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0428597 Certificate TypeCertificate of Analysis Date02/10/2023 Catalog Number122040100,122040250,122045000,122040025
Lot NumberA0410669 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number122040100,122040250,122045000,122040025
Lot NumberA0396557 Certificate TypeCertificate of Analysis Date21/11/2022 Catalog Number122040100,122040250,122045000,122040025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Imidazolidone hemihydrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.