missing translation for 'onlineSavingsMsg'
Learn More
2-Iodo-5-nitropyridine, 97%, Thermo Scientific Chemicals Product Code.: 11416190

2-Iodo-5-nitropyridine, 97%, Thermo Scientific Chemicals

Product Code. 11416190
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11416190

Brand: Thermo Scientific Alfa Aesar H27103.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28080-54-8
Molecular Formula C5H3IN2O2
Molecular Weight (g/mol) 249.995
MDL Number MFCD05863241
InChI Key SJXWHBQWFBHASX-UHFFFAOYSA-N
PubChem CID 819492
IUPAC Name 2-iodo-5-nitropyridine
SMILES C1=CC(=NC=C1[N+](=O)[O-])I

Specifications

Melting Point 163°C to 168°C
Quantity 5 g
Sensitivity Light sensitive
Synonym 6-iodo-3-nitropyridine, acmc-20alyk, pubchem6575, 5-nitro-2-iodopyridine, pyridine,2-iodo-5-nitro, 2-iodanyl-5-nitro-pyridine, pyridine, 2-iodo-5-nitro, 2-iodo-5-nitropyridine
Formula Weight 249.98
Percent Purity 97%
Chemical Name or Material 2-Iodo-5-nitropyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10223186 Certificate TypeCertificate of Analysis Date08/08/2023 Catalog NumberH27103.03,H27103.06
Lot Number10223186 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27103.03,H27103.06
Lot Number10136985 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27103.03,H27103.06
Lot Number10183908 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27103.03,H27103.06
Lot Number10139100 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27103.03,H27103.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Iodo-5-nitropyridine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.