missing translation for 'onlineSavingsMsg'
Learn More
2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals Product Code.: 11498386

2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals

Product Code. 11498386
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11498386

Brand: Thermo Scientific Alfa Aesar A15412.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.

Notes
Store in a cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-23-0
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00004066
InChI Key RZCJYMOBWVJQGV-UHFFFAOYSA-N
Synonym 2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapalShow MoreShow Less
PubChem CID 8422
ChEBI CHEBI:50397
IUPAC Name 2-naphthalen-2-yloxyacetic acid
SMILES C1=CC=C2C=C(C=CC2=C1)OCC(=O)O

Specifications

Melting Point 155°C to 157°C
Quantity 500 g
Beilstein 1074148
Merck Index 14,6397
Solubility Information Soluble in alcohol,ether,acetic acid and dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 2-Naphthoxyacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252373 Certificate TypeCertificate of Analysis Date13/09/2024 Catalog NumberA15412.22,A15412.36
Lot Number5002G26X Certificate TypeCertificate of Analysis Date19/10/2023 Catalog NumberA15412.22,A15412.36,A15412.0E
Lot NumberF8758A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15412.22,A15412.36
Lot NumberI6958A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15412.22,A15412.36
Lot Number5002G26X Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15412.22,A15412.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Naphthoxyacetic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.