missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitro-4-(trifluoromethyl)benzyl alcohol, 97+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used as important intermediates and reagents in organic synthesis. Also used as solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Used as important intermediates and reagents in organic synthesis. Also used as solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 63°C to 65°C |
Quantity | 1 g |
Formula Weight | 221.13 |
Percent Purity | ≥97% |
Chemical Name or Material | 2-Nitro-4-(trifluoromethyl)benzyl alcohol |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Nitro-4-(trifluoromethyl)benzyl alcohol, 97+%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction