missing translation for 'onlineSavingsMsg'
Learn More
2-Phenyl-4-penten-2-ol, 98%, Thermo Scientific™ Product Code.: 11439726

2-Phenyl-4-penten-2-ol, 98%, Thermo Scientific™

Product Code. 11439726
25 g
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Product Code. 11439726

Brand: Thermo Scientific Alfa Aesar B20248.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4743-74-2
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00060886
InChI Key BELRNEPYFJNSPN-UHFFFAOYSA-N
Synonym 2-phenyl-4-penten-2-ol, 2-phenyl 4-penten-2-ol, allyl methyl phenyl carbinol, phenyl-methyl-allyl-carbinol
PubChem CID 143761
IUPAC Name 2-phenylpent-4-en-2-ol
SMILES CC(CC=C)(C1=CC=CC=C1)O

Specifications

Density 0.99
Quantity 25 g
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 2-Phenyl-4-penten-2-ol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Phenyl-4-penten-2-ol, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.