missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex 2112 Com4-Fit Ear Plugs
Compact, pre-shaped design provides an ideal fit for users with smaller ear canals. Soft foam construction ensures a high level of comfort, even during long periods of use.
£5.35 - £33.80
Specifications
Size | Small |
---|---|
Model | 2112 Com4-Fit |
Corded | Uncorded |
Plug Shape | Oval |
Product Type | Ear Plugs |
Product Code | Brand | Packaging Type | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Price | Quantity & Availability | ||||||
17270342
|
uvex
2112131 |
Mini Box |
£5.35
15 pairs |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||||
15172443
|
uvex
2112004 |
Bagged in pairs |
£33.80
Each |
Estimated Shipment: 17-02-2025
Log in to see stock. |
||||||
17260342
|
uvex
2112096 |
Polybag |
£18.35
100 pairs |
Estimated Shipment: 24-02-2025
Log in to see stock. |
||||||
Description
- Compact, ergonomic, pre-shaped design sits comfortably on people with smaller ear canals
- Soft foam construction provides a high level of comfort, even after a very long period of use
- Closed skin prevents foreign bodies from penetrating the surface
- Ideal for transporting, and for smaller quantities
- Suitable for use at low noise levels
Specifications
Small | |
Uncorded | |
Ear Plugs | |
No |
2112 Com4-Fit | |
Oval | |
Single-use | |
No |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
uvex 2112 Com4-Fit Ear Plugs