missing translation for 'onlineSavingsMsg'
Learn More
2H-1,4-Benzoxazin-3(4H)-one, 99%, Thermo Scientific Chemicals Product Code.: 11496560

2H-1,4-Benzoxazin-3(4H)-one, 99%, Thermo Scientific Chemicals

Product Code. 11496560
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11496560

Brand: Thermo Scientific Alfa Aesar H27622.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5466-88-6
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00158536
InChI Key QRCGFTXRXYMJOS-UHFFFAOYSA-N
Synonym 2h-1,4-benzoxazin-3 4h-one, 2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3-one, 3-oxo-4h-benzo 1,4 oxazine, 3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3-ol, 2h-1,4-benzoxazin-3 4h-one 8ci 9ci, 4h-benzo 1,4 oxazin-3-one, 2h-1,4-benzoxazine-3 4h-one, 2h,4h-benzo e 1,4-oxazaperhydroin-3-oneShow MoreShow Less
PubChem CID 72757
IUPAC Name 4H-1,4-benzoxazin-3-one
SMILES C1C(=O)NC2=CC=CC=C2O1

Specifications

Melting Point 173°C to 175°C
Quantity 25 g
Formula Weight 149.15
Percent Purity 99%
Chemical Name or Material 2H-1,4-Benzoxazin-3(4H)-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248156 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog NumberH27622.06,H27622.14
Lot Number10247949 Certificate TypeCertificate of Analysis Date11/09/2023 Catalog NumberH27622.06,H27622.14
Lot Number10166802 Certificate TypeCertificate of Analysis Date01/05/2023 Catalog NumberH27622.14
Lot Number10166802 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27622.06,H27622.14
Lot Number10150852 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27622.06,H27622.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2H-1,4-Benzoxazin-3(4H)-one, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.