missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
50 g
100 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 10 g |
Formula Weight | 272.09 |
Percent Purity | 97% |
Chemical Name or Material | 3-(2, 6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction