missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

£40.00 - £524.00

Chemical Identifiers

CAS 39234-86-1
Molecular Formula C8H3ClF6O2S
Molecular Weight (g/mol) 312.61
MDL Number MFCD00014725
InChI Key BTRCVKADYDVSLI-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724
PubChem CID 520945
IUPAC Name 3,5-bis(trifluoromethyl)benzenesulfonyl chloride
SMILES C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11494043
View Documents
Thermo Scientific Alfa Aesar
A10300.03
1 g
£40.00
1g
Please to purchase this item. Need a web account? Register with us today!
11404053
View Documents
Thermo Scientific Alfa Aesar
A10300.06
5 g
£152.00
5g
Please to purchase this item. Need a web account? Register with us today!
11414053
View Documents
Thermo Scientific Alfa Aesar
A10300.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

39234-86-1
312.61
BTRCVKADYDVSLI-UHFFFAOYSA-N
520945
C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F
C8H3ClF6O2S
MFCD00014725
3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724
3,5-bis(trifluoromethyl)benzenesulfonyl chloride

Specifications

39234-86-1
C8H3ClF6O2S
UN3261
Moisture sensitive
BTRCVKADYDVSLI-UHFFFAOYSA-N
3,5-bis(trifluoromethyl)benzenesulfonyl chloride
520945
97%
34°C to 38°C
MFCD00014725
3621218
3,5-bis trifluoromethyl benzenesulfonyl chloride, 3,5-bis trifluoromethyl benzene-1-sulfonyl chloride, 3,5-bis trifluoromethyl benzenesulphonyl chloride, 3,5-di trifluoromethyl benzene-1-sulfonyl chloride, 3,5-ditrifluoromethylbenzenesulfonyl chloride, benzenesulfonyl chloride, 3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl phenylsulfonyl chloride, 3,5-bis-trifluoromethyl-benzenesulfonyl chloride, 3,5-di trifluoromethyl benzene sulfonyl chloride, pubchem2724
C1=C(C=C(C=C1C(F)(F)F)S(=O)(=O)Cl)C(F)(F)F
312.61
312.61
3,5-Bis (trifluoromethyl)benzenesulfonyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 254-371-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only