missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals Product Code.: 10539823

3,5-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 10539823
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10539823

Brand: Thermo Scientific Acros 116000050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-06-9
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00002525
InChI Key UMVOQQDNEYOJOK-UHFFFAOYSA-N
Synonym mesitylenic acid, benzoic acid, 3,5-dimethyl, unii-ed8av34n0y, 3,5-dimethyl benzoic acid, 3,5-dimethyl-benzoic acid, ed8av34n0y, 3,5-dimethylbenzoicacid, pubchem15440, 3,5-dimethylbezoic acid, 3,5 dimethylbenzoic acidShow MoreShow Less
PubChem CID 10356
ChEBI CHEBI:64821
IUPAC Name 3,5-dimethylbenzoic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C

Specifications

Melting Point 169°C to 172°C
CAS Min % 98.5
Color White to Yellow
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 99%
Linear Formula (CH3)2C6H3CO2H
Beilstein 09, 536
Solubility Information Solubility in water: slightly soluble
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 3, 5-Dimethylbenzoic acid, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443601 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number116001000,116000250,116000050
Lot NumberA0443601 Certificate TypeCertificate of Analysis Date28/06/2022 Catalog Number
Lot NumberA0435328 Certificate TypeCertificate of Analysis Date17/01/2022 Catalog Number116001000,116000250,116000050
Lot NumberA0340853 Certificate TypeCertificate of Analysis Date03/05/2021 Catalog Number116001000,116000250,116000050
Lot NumberA0423737 Certificate TypeCertificate of Analysis Date26/10/2020 Catalog Number116001000,116000250,116000050
compliance-icons
Product Identifier
  • 3,5-Dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dimethylbenzoic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.