missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-2,6-dibromopyridine, 97%, Thermo Scientific™ Product Code.: 11946491

3-Amino-2,6-dibromopyridine, 97%, Thermo Scientific™

Product Code. 11946491
5 g
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy

Product Code. 11946491

Brand: Thermo Scientific Acros 447640050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 39856-57-0
Melting Point 147°C to 148°C
Assay Percent Range 96% min. (GC)
Molecular Formula C5H4Br2N2
Quantity 5 g
Synonym 3-amino-2,6-dibromopyridine, 2,6-dibromo-3-pyridinamine, 2,6-dibromo-pyridin-3-ylamine, 3-pyridinamine, 2,6-dibromo, 2,6-dibromo-3-pyridylamine, 2,6-dibromo-3-aminopyridine, zlchem 949, pubchem9809, acmc-209j8e, ksc495i3f
InChI Key CRTOIQFRVBJJRI-UHFFFAOYSA-N
SMILES C1=CC(=NC(=C1N)Br)Br
IUPAC Name 2,6-dibromopyridin-3-amine
Molecular Weight (g/mol) 251.91
PubChem CID 817678
Formula Weight 251.91
Percent Purity 97%
Chemical Name or Material 3-Amino-2, 6-dibromopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-2,6-dibromopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-2,6-dibromopyridine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.