missing translation for 'onlineSavingsMsg'
Learn More
3-Bromo-5-nitro-1H-indazole, 95%, Thermo Scientific™ Product Code.: 15477847

3-Bromo-5-nitro-1H-indazole, 95%, Thermo Scientific™

Product Code. 15477847
10 g
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy

Product Code. 15477847

Brand: Thermo Scientific Alfa Aesar H37979.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67400-25-3
Molecular Formula C7H4BrN3O2
Molecular Weight (g/mol) 242.03
MDL Number MFCD07367964
InChI Key SYTGRKBQTUKOAI-UHFFFAOYSA-N
Synonym 3-bromo-5-nitro-1h-indazole, 3-bromo-5-nitroindazole, 5-nitro-3-bromoindazole, 1h-indazole, 3-bromo-5-nitro, 1h-indazole,3-bromo-5-nitro, 5-nitro-3-bromo-indazole, pubchem20224, 3-bromo-5-nitro1h-indazole, ksc497g0j, 3-bromo-5-nitro 1h-indazoleShow MoreShow Less
PubChem CID 640614
IUPAC Name 3-bromo-5-nitro-2H-indazole
SMILES [O-][N+](=O)C1=CC2=C(Br)NN=C2C=C1

Specifications

Melting Point 227°C to 229°C
Color Brown
Quantity 10 g
Formula Weight 242.03
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 3-Bromo-5-nitro-1H-indazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10178006 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-5-nitro-1H-indazole, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.