missing translation for 'onlineSavingsMsg'
Learn More
3-Bromooxetane, 95%, Thermo Scientific Chemicals Product Code.: 15463908

3-Bromooxetane, 95%, Thermo Scientific Chemicals

Product Code. 15463908
500 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15463908

Brand: Thermo Scientific Alfa Aesar H57437.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39267-79-3
Molecular Formula C3H5BrO
Molecular Weight (g/mol) 136.98
MDL Number MFCD08544402
InChI Key SZTIZZFKWQWSSP-UHFFFAOYSA-N
Synonym 3-bromo-oxetane, 3-bromo oxetane, 3-bromanyloxetane, 3-bromooxetane, acmc-209j4f, 2-bromo-1,3-epoxypropane, 5-n,n-dimethylamino-2-hydroxybenzaldehydeShow MoreShow Less
PubChem CID 16244493
IUPAC Name 3-bromooxetane
SMILES BrC1COC1

Specifications

Density 1.793
Boiling Point 131°C
Flash Point 48°C (118°F)
Quantity 500 mg
UN Number UN1993
Formula Weight 136.98
Percent Purity 95%
Chemical Name or Material 3-Bromooxetane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR22E066 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57437.ME,H57437.MD
Lot NumberD06Y018 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57437.ME,H57437.MD
Lot NumberT27E040 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57437.ME,H57437.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromooxetane, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.