missing translation for 'onlineSavingsMsg'
Learn More
(3-Methylphenyl)methanethiol, 95%, Thermo Scientific™ Product Code.: 10496482

(3-Methylphenyl)methanethiol, 95%, Thermo Scientific™

Product Code. 10496482
1 g
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 10496482

Brand: Thermo Scientific Maybridge TL00469DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25697-56-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00022075
InChI Key PNAZUQUHTIOEHF-UHFFFAOYSA-N
PubChem CID 2801207
IUPAC Name (3-methylphenyl)methanethiol
SMILES CC1=CC(=CC=C1)CS

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula CH3C6H4CH2SH
Quantity 1 g
Synonym 3-methylphenyl methanethiol, 3-methylbenzyl mercaptan, m-methylbenzyl mercaptan, 3-methylbenzenemethanethiol, m-methylbenzylmercaptan, acmc-1cq7k, 3-methyl-benzenemethanethiol, benzenemethanethiol,3-methyl, benzenemethanethiol, 3-methyl, 3-methylphenyl methane-1-thiol
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material (3-Methylphenyl)methanethiol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(3-Methylphenyl)methanethiol, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.