Learn More
Applied Biosystems™ Sequencing Standard, BigDye™ Terminator v3.1, for 3500/SeqStudio™ Flex
The DNA contained in the preparation has been lyophilized to maximize stability.The two hydrolytic enzymes used in this kit, shrimp alkaline phosphatase and exonuclease I, effectively remove excess dNTPs and primers.
Brand: Applied Biosystems™ 4404312
272.64 GBP valid until 2025-12-31
Use promo code "24365" to get your promotional price.
Includes: Kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye® Terminator v3.1. Each tube can perform one spectral calibration/performance check and/or one control sequencing run on the Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers.
Description
The BigDye Terminator v3.1 sequencing standard kit contains four tubes, each of which contains enough DNA of a known 1200 base pair sequence prepared with BigDye Terminator v3.1 to perform one spectral calibration/performance check and/or one control sequencing run on Applied Biosystems 3500 and SeqStudio Flex series genetic analyzers. The DNA contained in the preparation has been lyophilized to maximize stability.Order Info
4 Tubes

Specifications
This kit contains four tubes of DNA of a known 1200 base pair sequence prepared with BigDye Terminator v3.1. Each tube can perform one spectral calibration⁄performance check and⁄or one control sequencing run on Applied Biosystems 3500 or SeqStudio Flex series genetic analyzer. Store kit at -15 to -25°C. | |
Sequencing | |
BigDye™ Terminator, BigDye™ | |
4 Tubes |
3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers | |
ROX, TAMRA™, Rhodamine 110, Rhodamine 6G | |
Sequencing Standards | |
Dry Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2831743 | Certificate of Analysis | 31/01/2025 | 4404312 |
2815473 | Certificate of Analysis | 29/01/2025 | 4404312 |
2809991 | Certificate of Analysis | 29/01/2025 | 4404312 |
2809995 | Certificate of Analysis | 29/01/2025 | 4404312 |
2789506 | Certificate of Analysis | 29/01/2025 | 4404312 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.