Learn More
3M™ PAPR Battery Charger Cradle
TR-340 Battery Charger Cradle provides an easy-to-use interface between all TR-300 batteries and all TR-900 Series power supplies.
Brand:  3M™ 7000103971
126.41 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11733906
  • £202.00 / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement part for the TR-341 and TR-344 battery charger kits
  • 3M Catalog# TR-340
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Single-Unit Charging Station
Versaflo™ TR-300 and Speedglas™ TR-300 PAPR Systems
One
Provides easy-to-use interface between all TR-300 batteries and all TR-900 series power supplies
TR-340
1 Case
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ PAPR Battery Charger Cradle > Battery charger cradle