missing translation for 'onlineSavingsMsg'
Learn More

4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Thermo Scientific™

£17.30 - £164.00

Chemical Identifiers

CAS 160068-88-2
Molecular Formula C10H11BO3
Molecular Weight (g/mol) 190.005
MDL Number MFCD04039026
InChI Key AMQBUMNOSGJNQL-UHFFFAOYSA-N
Synonym 4-1,3,2-dioxaborinan-2-yl benzaldehyde, 4-formylbenzeneboronic acid, propane-1,3-diol cyclic ester, 4-formylbenzeneboronic acid,propane-1,3-diol cyclic ester, 4-1,3,2-dioxaboran-2-yl benzaldehyde, 2-4-formylphenyl-1,3,2-dioxaborinane
PubChem CID 2794711
IUPAC Name 4-(1,3,2-dioxaborinan-2-yl)benzaldehyde
SMILES B1(OCCCO1)C2=CC=C(C=C2)C=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10423573
SDS View Documents
Thermo Scientific Maybridge
MO01234CB
250 mg
£17.30
250mg
Please to purchase this item. Need a web account? Register with us today!
10600105
SDS View Documents
Thermo Scientific Maybridge
MO01234DA
1 g
£35.70
1g
Please to purchase this item. Need a web account? Register with us today!
10699505
SDS View Documents
Thermo Scientific Maybridge
MO01234DE
5 g
£164.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(1,3,2-Dioxaborinan-2-yl)benzaldehyde, ≥97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.