missing translation for 'onlineSavingsMsg'
Learn More
4,16-Dibromo[2.2]paracyclophane, 98%, Thermo Scientific Chemicals Product Code.: 15423307

4,16-Dibromo[2.2]paracyclophane, 98%, Thermo Scientific Chemicals

Product Code. 15423307
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15423307

Brand: Thermo Scientific Alfa Aesar B25745.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96392-77-7
Molecular Formula C16H14Br2
MDL Number MFCD09953451

Specifications

Melting Point 249°C to 252°C
Quantity 5 g
Formula Weight 366.1
Percent Purity 98%
Chemical Name or Material 4,16-Dibromo[2.2 ]paracyclophane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,16-Dibromo[2.2]paracyclophane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.