missing translation for 'onlineSavingsMsg'
Learn More
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%, Thermo Scientific Chemicals Product Code.: 10711254

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%, Thermo Scientific Chemicals

Product Code. 10711254
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10711254

Brand: Thermo Scientific Acros 384640010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552846-17-0
Molecular Formula C14H23BN2O4
Molecular Weight (g/mol) 294.16
MDL Number MFCD05663873
InChI Key IPISOFJLWYBCAV-UHFFFAOYSA-N
Synonym 1-boc-pyrazole-4-boronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole-1-carboxylate, 1-boc-4-pyrazoleboronic acid pinacol ester, 1-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole-1-carboxylate, n-boc-pyrazole-4-boronic acid pinacol ester, 1-tert-butoxycarbonyl-1h-pyrazole-4-boronic acid, pinacol ester, n-boc-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1-tert-butoxycarbonyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-tert-butoxycarbonyl-4-1h-pyrazoleboronic acidShow MoreShow Less
PubChem CID 16217654
IUPAC Name tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 82°C to 89°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 294.16
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-butyl ester
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0383962 Certificate TypeCertificate of Analysis Date04/05/2022 Catalog Number384640050,384640010
compliance-icons
Product Identifier
  • 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic acid tert-butyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-carboxylic Acid Tert-Butyl Ester, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.