missing translation for 'onlineSavingsMsg'
Learn More
4-Amino-1,8-naphthalimide, 95%, Thermo Scientific Chemicals Product Code.: 15476339

4-Amino-1,8-naphthalimide, 95%, Thermo Scientific Chemicals

Product Code. 15476339
1 g
Click to view available options
Quantity:
100 mg
1 g
This item is not returnable. View return policy

Product Code. 15476339

Brand: Thermo Scientific Alfa Aesar J64358.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1742-95-6
Molecular Formula C12H8N2O2
Molecular Weight (g/mol) 212.21
MDL Number MFCD00006921
InChI Key SSMIFVHARFVINF-UHFFFAOYSA-N
Synonym 4-amino-1,8-naphthalimide, 4-aminonaphthalimide, 6-amino-benzo de isoquinoline-1,3-dione, 6-amino-1h-benzo de isoquinoline-1,3 2h-dione, 4-aminonaphthalene-1,8-dicarboximide, naphthalimide, 4-amino, dfp 1, 1h-benz de isoquinoline-1,3 2h-dione, 6-amino, 6-aminobenzo de isoquinoline-1,3-dione, parp inhibitor v, 4-aniShow MoreShow Less
PubChem CID 1720
ChEBI CHEBI:40071
SMILES NC1=C2C=CC=C3C(=O)NC(=O)C(C=C1)=C23

Specifications

Melting Point >360°C
Color Orange
Density 1.105
Quantity 1 g
Solubility Information Soluble in DMSO
IUPAC Name 10-amino-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione
Formula Weight 212.21
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Amino-1,8-naphthalimide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY30G035 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ64358.MC,J64358.03
Lot NumberS26K026 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog NumberJ64358.MC,J64358.03
Lot NumberT13I015 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog NumberJ64358.MC,J64358.03
Lot NumberT09G007 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ64358.MC,J64358.03
Lot NumberP14C077 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberJ64358.MC,J64358.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Amino-1,8-naphthalimide, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.