missing translation for 'onlineSavingsMsg'
Learn More
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals Product Code.: 11394404

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Product Code. 11394404
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11394404

Brand: Thermo Scientific Alfa Aesar L00982.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

Solubility
Soluble in Dimethyl sulfoxide. (DMSO)

Notes
Store at -20°C. Avoid contact with heat. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1750-12-5
Molecular Formula C2H6N6S
Molecular Weight (g/mol) 146.172
MDL Number MFCD00003098
InChI Key RDIMQHBOTMWMJA-UHFFFAOYSA-N
Synonym 4-amino-3-hydrazino-5-mercapto-1,2,4-triazole, purpald, 4-amino-3-hydrazino-1,2,4-triazol-5-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, hydrazone, purpald r, 4-amino-5-hydrazino-1,2,4-triazole-3-thiol, 1,2,4-triazolidin-3-one, 4-amino-5-thioxo-, 3-hydrazone, 4-amino-5-hydrazinyl-4h-1,2,4-triazole-3-thiol, 3h-1,2,4-triazole-3-thione, 4-amino-5-hydrazinyl-2,4-dihydro, 4-amino-5-hydrazinyl-1,2,4-triazole-3-thiolShow MoreShow Less
PubChem CID 2723946
IUPAC Name 4-amino-3-hydrazinyl-1H-1,2,4-triazole-5-thione
SMILES C1(=S)NN=C(N1N)NN

Specifications

Melting Point ∽220°C (decomposition)
Quantity 25 g
UN Number UN1325
Beilstein 134867
Solubility Information Soluble in Dimethyl sulfoxide. (DMSO)
Formula Weight 146.17
Percent Purity ≥99%
Chemical Name or Material 4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250176 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog NumberL00982.06,L00982.14
Lot Number10243829 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberL00982.06,L00982.14
Lot Number10180576 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00982.06,L00982.14
Lot Number10237033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00982.06,L00982.14
Lot Number10175500 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL00982.06,L00982.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.