Learn More
4-Aminobutyric acid, 99+%, Thermo Scientific Chemicals
4-Aminobutyric acid, 99+%, CAS # 56-12-2, also known as γ-Aminobutyric acid, is an amino acid that shows inhibitory activities against neurotransmitters, including GABA.
£23.52 - £685.41
Chemical Identifiers
CAS | 56-12-2 |
---|---|
Molecular Formula | C4H9NO2 |
Molecular Weight (g/mol) | 103.12 |
MDL Number | MFCD00008226 |
InChI Key | BTCSSZJGUNDROE-UHFFFAOYSA-N |
Synonym | 4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino |
PubChem CID | 119 |
ChEBI | CHEBI:16865 |
IUPAC Name | 4-aminobutanoic acid |
SMILES | C(CC(=O)O)CN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10775931
|
Thermo Scientific Acros
103280250 |
25 g | Plastic Bottle |
£23.52
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10244030
|
Thermo Scientific Acros
103281000 |
100 g | Plastic Bottle |
£59.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10518470
|
Thermo Scientific Acros
103285000 |
500 g | Plastic Bottle |
£173.87
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15129153
|
Thermo Scientific Acros
103280025 |
2.5 kg | Plastic Bottle |
£685.41
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• 4-Aminobutyric acid is a γ-amino acid and monocarboxylic acid derived from butyric acid
• It is an amino acid that shows inhibitory activities against neurotransmitters, including GABA, and acts by binding GABA-A receptor and inhibiting the activity of signal-receiving neurons
• This compound, converted from neurotransmitter glutamate, is essential to regulate neuronal excitability
• It binds to GABA-A and GABA-B receptors, causing ion channel opening, hyperpolarization, and eventually inhibition of neurotransmission
Applications
• 4-Aminobutyric acid has been used in the preparation of GABA–BSA (bovine serum albumin)–GA (glutaraldehyde) conjugate, which is used as a control for immunohistochemical experiments
• It is used, in vivo, to detect GABA neurotransmitter by magnetic resonance spectroscopy
• This compound increases the production of short-chain fatty acids and decreases pH values in mouse colon
• Some in vivo experiments, using rats, mice, and pigs, demonstrated that 4-Aminobutyric acid can modulate intestinal fluid secretion, blood flow and ulceration
Chemical Identifiers
56-12-2 | |
103.12 | |
BTCSSZJGUNDROE-UHFFFAOYSA-N | |
119 | |
4-aminobutanoic acid |
C4H9NO2 | |
MFCD00008226 | |
4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino | |
CHEBI:16865 | |
C(CC(=O)O)CN |
Specifications
197°C to 204°C | |
99.0 | |
White | |
99+% | |
C4H9NO2 | |
MFCD00008226 | |
15,425 | |
4-aminobutyric acid, gaba, gamma-aminobutyric acid, piperidic acid, piperidinic acid, aminalon, gaballon, gammalon, mielogen, butanoic acid, 4-amino | |
BTCSSZJGUNDROE-UHFFFAOYSA-N | |
4-aminobutanoic acid | |
119 | |
103.12 | |
Crystalline Powder |
56-12-2 | |
100.0 | |
Authentic | |
0.5% max. | |
H2N(CH2)3CO2H | |
04,413 | |
95% min. (c=10, 2N HCl, 430nm) 1cm cell | |
Solubility in water: very soluble. Other solubilities: slightly soluble in common solvents | |
C(CC(=O)O)CN | |
103.12 | |
CHEBI:16865 | |
≥99% | |
4-Aminobutyric acid, 99+% |
Safety and Handling
missing translation for 'einecsNumber' : 200-258-6
missing translation for 'rtecsNumber' : ES6300000
missing translation for 'tsca' : TSCA
- 4-Aminobutyric acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.